Läs mer
Invitrogen™ Histone H1T Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566793
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: RATTPKTVRSG RKAKGAKGKQ QQKSPVKARA SKSKLTQHHE VNVRKATSKK Highest antigen sequence identity to the following orthologs - mouse 48%, rat 48%.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.
Specifikationer
| Histone H1T | |
| Polyclonal | |
| Unconjugated | |
| H1-6 | |
| dJ221C16.2; H1 histone family, member T (testis-specific); H1.6; H1ft; H1T; HIST1H1T; histone 1, H1t; histone cluster 1, H1t; histone H1t; Testicular H1 histone | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P22492 | |
| H1-6 | |
| Recombinant Human Histone H1T. Recombinant protein control fragment (Product #RP-107450). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |