missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2A.z Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16441-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.Specifications
Histone H2A.z | |
Polyclonal | |
Unconjugated | |
H2AFZ | |
H2A histone family, member Z, H2A.z, H2A/z, H2AFZ, H2AZ, Histone H2A.Z | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3015, 51788, 58940 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0C0S5, P0C0S6, P0C0S7 | |
H2AFZ | |
Histone H2A.z Fusion Protein Ag9762 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |