Learn More
Epredia™ HistoScreen™ Cassettes
Product Code. 16333926 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16333926 Blue Pack of 1000
16323926 Beige Pack of 1000
16353926 Gray Pack of 1000
16343926 Green Pack of 1000
16363926 Lavender Pack of 1000
16373926 Light Green Pack of 1000
16383926 Lilac Pack of 1000
16393926 Orange Pack of 1000
16303936 Pink Pack of 1000
16313936 Tan Pack of 1000
16323936 White Pack of 1000
16333936 Yellow Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 16333926 Supplier Epredia™ Supplier No. B851000BL
  • ONLINE EXCLUSIVE PRICE
    6750.00 SEK
    5103.68 SEK / Paket med 1000
    Save 1646.32 SEK
    24% Av
 Restricted Item
I lager
For very small biopsy tissue samples with a back-edge lid hinge. Eliminates the need for sponges or biopsy bags.
  • Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
  • Grossing, Processing and Embedding – cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
  • Precision engineering for solid connection to microtomes.
  • Cassette surface texture optimizes scanability for archiving.
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Mesh
Closure Included Separate lids included
Quantity 1000
Closure Type Back Hinge
Color Blue
Product Type HistoScreen™ Cassette