Läs mer
Epredia™ HistoScreen™ Cassettes in Tube Packs, aqua
Handla allt Epredia Produkter
Färg:
Blått
Grå
Grön
Gult
Lila
Orange
Rosa
Solbränna
Turkos
Vitt
Förpackningsstorlek:
Paket med 750
Varje
Beskrivning
- Saves time, offers the greatest specimen security, and provides ease of use
- Provides an inner chamber lined with a “see-through” mesh for easy identification of specimens
- Eliminates the need for messy sponges or lens paper
- Ensures your tissues are safely processed with cassettes which will not pop open during processing
Specifikationer
Specifikationer
| Por stil | Mesh Pore Insert |
| Färg | Turkos |
| Produkttyp | HistoScreen™ Cassette |