missing translation for 'onlineSavingsMsg'
Learn More

Simport™ HistoTainer™ I Prefilled Specimen Containers

These tamper-evident specimen containers are 50% prefilled with 10% neutral buffered Formalin.

Brand:  Simport™ M960-60FMA

Additional Details : Weight : 4.72000kg

Product Code. 15817114

  • 436.00 SEK / Pack of 96

Estimated Shipment: 01-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Container made of polypropylene, closure made of polyethylene
  • Especially designed for the collection, transport and storage of histology specimens
  • Shatter-resistant, eliminating most problems of leakage and evaporation
  • Magenta lids are ribbed for easy opening when hands are wet or gloved
  • Jars are stackable for easy, safe storage and translucent to allow specimens to be viewed without opening
  • Innovative tamper-evident screw cap
  • Formalin is enhanced by a buffering capacity optimizing histological results by light microscopy and immunohistochemistry
Specifications

Specifications

60 mL
Magenta
Polyethylene
Neutral buffered formalin
Polypropylene
Screw Cap
Histology specimens
24/Tray, 96/Cs
Videos
SDS
Documents
Certificates