missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
HIV-1 Gag p24 Antibody (ND1), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-473JF549
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifikationer
| HIV-1 Gag p24 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| gag | |
| HIV-1 Gag p24 C-terminal peptide. | |
| 0.1 mL | |
| 155030 | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| ND1 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24 | |
| Mouse | |
| Ion exchange chromatography | |
| Primary | |
| The antibody reacts with HIV-1 Gag p24). Specificity was verified by Western Blotting in reducing conditions with Sanofi Pasteur NEW LAV-BLOT I kit. Epitope was reported to be located within the amino acids 51-110. | |
| Store at 4°C in the dark. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering