Läs mer
Tocris Bioscience™ HJC 0350
GREENER_CHOICE
Produktkod. 30001559
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001559 10 mg
30001579 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001559 Leverantör Tocris Bioscience™ Leverantörsnummer 4844/10
Selective Epac2 inhibitor
Selective Epac2 inhibitor (IC50 = 0.3 μM). Displays no effect on Epac1. Blocks stimulation of the Epac2-FL FRET sensor in HEK293 cells.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 885434-70-8
Molekylformel C15H19NO2S
Specifikationer
Kemiskt namn eller material EPAC Inhibitors
Kvantitet 10 mg
Formel vikt 277.38
Renhet >98%
Molekylvikt (g/mol) 277.382