missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15656-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.Specifications
HK1 | |
Polyclonal | |
Unconjugated | |
HK1 | |
Hk1 | |
HK1 Fusion Protein Ag8008 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05708, P17710, P19367 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15275, 25058, 3098 | |
-20°C | |
Liquid |