missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ HK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597828
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
HK2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
In vertebrates there are four major glucose-phosphorylating isoenzymes, designated hexokinase I, II, III, and IV. Hexokinase activity is involved in the first step in several metabolic pathways including phosphorylation of glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. Hexokinase is an allosteric enzyme inhibited by its product GLC-6-P. Hexokinase 2 is the predominant hexokinase isozyme expressed in insulin-responsive tissues such as skeletal muscle. Expression of this gene is insulin-responsive, and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells.
Specifikationer
| HK2 | |
| Polyclonal | |
| Unconjugated | |
| Hk2 | |
| AI642394; DKFZp686M1669; Hexokinase; hexokinase 2; hexokinase II; hexokinase type II; Hexokinase-2; hexokinase-2 muscle; hexokinase-2, muscle; Hexokinase-B; HK II; Hk2; HKII; HXK2; muscle form hexokinase; OTTHUMP00000202738 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 3099 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P52789 | |
| Hk2 | |
| Recombinant Human Hexokinase-2 protein (334-526AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering