Learn More
HLA-ABC Monoclonal Antibody (W6/32), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
2890.00 SEK
Specifications
Antigen | HLA-ABC |
---|---|
Clone | W6/32 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15558706
|
Affymetrix eBioscience
25-9983-42 |
100 Tests |
2890.00 SEK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The W6/32 monoclonal antibody reacts with the human major histocompatibility complex (MHC) class I, HLA-A, B, C. MHC class I antigens associated with beta 2-microglobulin are expressed by all human nucleated cells and are central in cell-mediated immune response and tumor surveillance. W6/32 mAb recognizes a non-polymorphic epitope shared among products of the HLA-A, B, and C loci and immunoprecipitates both 43 kDa and 11-12 kDa chains. Crossreactivity is also seen in baboon, rhesus and cynomolgus monkey. Applications Reported: This W6/32 antibody has been reported for use in flow cytometric analysis. Applications Tested: This W6/32 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
HLA-ABC Monoclonal antibody specifically detects HLA-ABC in Human samples. It is validated for Flow Cytometry.Specifications
HLA-ABC | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
W6/32 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.