Learn More
HLA-BC Monoclonal Antibody (B1.23.2), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1048.00 SEK - 2970.00 SEK
Specifications
Antigen | HLA-BC |
---|---|
Clone | B1.23.2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15518166
|
Affymetrix eBioscience
17-5935-42 |
100 Tests |
2970.00 SEK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15508166
|
Affymetrix eBioscience
17-5935-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This B1.23.2 monoclonal antibody recognizes the human Human Leukocyte Antigens (HLA) B and C. Along with HLA-A, these proteins correspond to MHC Class I in humans. HLA-B and HLA-C are expressed on nearly all cell types and heterodimerize with beta-2 microglobulin at the cell surface. These molecular complexes play a critical role in the presentation of endoplasmic reticulum-derived peptides to immune cells. Applications Reported: This B1.23.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B1.23.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
HLA-BC Monoclonal antibody specifically detects HLA-BC in Human samples. It is validated for Flow Cytometry.Specifications
HLA-BC | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P01889 | |
HLA-B | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
HLA-B |
B1.23.2 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
3106 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.