Learn More
HLA-DOB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553095
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KADCYFTNGT EKVQFVVRFI FNLEEYVRFD SDVGMFVALI KLGQPDAEQW NSRLDLLERS RQAVDGVCRH NYRLGAPFTV GRKVQPEVTV YPERTPLLHQ HNLLHCSVTG FYPGDIKIKW FLNGQEERAG VMSTGPIRNG D Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.
HLA-DOB Polyclonal antibody specifically detects HLA-DOB in Human samples. It is validated for Immunohistochemistry.Specifications
HLA-DOB | |
Polyclonal | |
Unconjugated | |
HLA-DOB | |
DOB; HLA class II histocompatibility antigen, DO beta chain; HLA-DO beta; HLA-DOB; major histocompatibility complex, class II, DO beta; MHC class II antigen DOB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3112 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P13765 | |
HLA-DOB | |
Recombinant protein corresponding to Human HLA-DOB | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only