missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR Mouse anti-Canine, Cynomolgus Monkey, Human, Non-human primate, Rhesus Monkey, eFluor® 450, Clone: L243, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-9952-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15579846

  • 1250.00 SEK / 25 tests


Estimated Shipment
04-01-2021
Add to basket

Description

Description

HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response.
Specifications

Specifications

HLA-DR
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01903
XXbac-BPG161M6.1, DRB1, DRw10, FLJ75017, FLJ76359, HLA-DR1B, HLA-DRB, HLA-DRB1*, SS1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey
Flow Cytometry
L243
eFluor™ 450
HLA-DRA
Liquid
HLA-DRA
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3122
Videos
SDS
Documents

For Research Use Only.