missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-G Mouse anti-Human, Clone: 1E5A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66447-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail.Specifications
HLA-G | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17693 | |
HLA-G | |
HLA-G Fusion Protein Ag10839 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1E5A10 | |
Unconjugated | |
HLA-G | |
HLA 6.0, HLA G, HLA G antigen, HLAG, HLA-G, MHC class I antigen G, MHC G | |
Mouse | |
Protein A | |
RUO | |
3135 | |
-20°C | |
Liquid |