Läs mer
Tocris Bioscience™ HLI 373
GREENER_CHOICE
Produktkod. 18034712
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18034712 10 mg
18084292 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18034712 Leverantör Tocris Bioscience™ Leverantörsnummer 3503/10
Hdm2 inhibitor; activates p53-dependent transcription
Inhibitor of Hdm2 ubiquitin ligase (E3). Blocks Hdm2-mediated ubiquitylation and proteasomal degradation of p53; activates p53-dependent transcription. Induces apoptosis in several tumor cell lines that express wild-type p53 such as LOX-IMVI, A549, HT1080 and U2OS.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1782531-99-0
Specifikationer
Kemiskt namn eller material HLI 373
Kvantitet 10 mg
Molekylformel C18H25Cl2N5O2
Formel vikt 414.33
Renhet >99%
Molekylvikt (g/mol) 414.33