missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Description
HLX1 Polyclonal antibody specifically detects HLX1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | HLX1 |
| Användningsområden | Western Blot, Immunoprecipitation |
| Klassificering | Polyclonal |
| Konjugera | PE |
| Utspädning | Western Blot, Immunoprecipitation |
| Formulering | PBS |
| Gene Alias | H2.0 (Drosophila)-like homeo box 1, H2.0-like homeo box 1, H2.0-like homeo box-1, H2.0-like homeobox, H2.0-like homeobox 1, H2.0-like homeobox protein, HB24, HLX1H2.0-like homeobox 1 (Drosophila), Homeobox protein HB24, Homeobox protein HLX1 |
| Värd art | Rabbit |
| Immunogen | Maps to a region between residue 375 and 425 of human H2.0-like Homeobox using the numbering given in entry NP_068777.1 (GeneID 3142). |
| Reningsmetod | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?