Learn More
Epredia™ HM 650V Vibrating Blade Microtome Specimen Stage F (Flat)for medium to large specimens or tissue (from 10mm to 45mm)
Use these Epredia™ HM 650V Vibrating Blade Microtome Accessories with the Epredia™ HM650V Vibrating Blade Microtome.
Brand:  Epredia™ 760320
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12057249
  • 4940.00 SEK / Each
Estimated Shipment: 15-12-2021
Description
Description
Standard buffer tray:
  • Hold stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
Cooling element for buffer solution:
  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
Large-field magnifier:
  • Magnification: 2.5x
  • Ability to move, swivel, and displace for optimal viewing position
  • Adapter and illumination included
StemiScope™ with optics and mount:
  • Eyepieces: 10x
  • Supplementary lens: 0.63x
  • Zoom magnifying range: 4.1 to 31.5x
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
Swan-neck light guide:
  • 3.5mm fiber bundle
  • 50mm long swan-neck light guide
  • 750mm long flexible light guide
  • High light intensity
  • Adapter for the KL200 (760317) included
Specifications
Specifications
Light Guide
HM 650V
Swan-Neck Light Guide
Swan-neck light guide