Learn More
HMBS Mouse anti-Human, Unconjugated, Clone: 1G12H3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67456-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840096
  • 1236.20 SEK / 20µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The HMBS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HMBS. This antibody recognizes human antigen. The HMBS antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
HMBS
Monoclonal
1000 μg/mL
Mouse
Primary
Human
ELISA, Immunohistochemistry, Western Blot
1G12H3
Unconjugated
20 μL
3145
IgG1