missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HMGN1 (Human) Recombinant Protein
Human HMGN1 full-length ORF ( AAH23984, 1 a.a. - 100 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003150-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPKRKVSSTEGAAKEEPKRRSARLSAKPPAKVEAKPKKAAAKDKSSDKKVQTKGKRGAKGKQAEVANQETKEDLPAENGETKTEESPASDEAGEKEAKSD
Specifications
AAH23984 | |
high-mobility group nucleosome binding domain 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HMGN1 | |
GST |
3150 | |
Wheat germ expression system | |
10 μg | |
FLJ27265, FLJ31471, HMG14, MGC104230, MGC117425 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |