missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGN4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HMGN4 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838933
|
Proteintech
11686-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828933
|
Proteintech
11686-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HMGN4, also named as High mobility group nucleosome-binding domain-containing protein 4, is a 90 amino acid protein, which belongs to the HMGN family. HMGN4 as a DNA-binding protein localizes in the nucleus. The calculated molecular weight of HMGN4 is 10 kDa, but the modified HMGN4 protein is about 15 kDa.Specifications
HMGN4 | |
Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O00479 | |
10473 | |
HMGN4 Fusion Protein Ag2298 | |
Primary | |
-20°C | |
HMGN4 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HMG17L3, HMGN4, NHC | |
HMGN4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |