missing translation for 'onlineSavingsMsg'
Learn More

HMGN4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11686-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828933

  • 4721.67 SEK / 150µL
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HMGN4, also named as High mobility group nucleosome-binding domain-containing protein 4, is a 90 amino acid protein, which belongs to the HMGN family. HMGN4 as a DNA-binding protein localizes in the nucleus. The calculated molecular weight of HMGN4 is 10 kDa, but the modified HMGN4 protein is about 15 kDa.
Specifications

Specifications

HMGN4
Polyclonal
Unconjugated
HMGN4
HMG17L3, HMGN4, NHC
Rabbit
Antigen Affinity Chromatography
RUO
10473
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00479
HMGN4
HMGN4 Fusion Protein Ag2298
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers