Learn More
HMHA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554087
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QRAEVVVQVP YLEAGEAVVY PLQEAAADGC RESRVVSNDS DSDLEEASEL LSSSEASALG HLSFLEQQQS EASLEVASGS HSGSEEQLEA TAREDGDGDE DGPAQQLSGF NTNQSNNV Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 56%.
HMHA1 Polyclonal antibody specifically detects HMHA1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
HMHA1 | |
Polyclonal | |
Unconjugated | |
ARHGAP45 | |
6330406L22Rik; ARHGAP45; AW539505; Ha-1; histocompatibility (minor) HA-1; HLA-HA1; Hmha1; KIAA0223; mHag HA-1; Minor histocompatibility antigen HA-1; minor histocompatibility protein HA-1; minor histocompatibility protein HA-1; rho GTPase-activating protein 45; RGD1308662; Rho GTPase activating protein 45; rho GTPase-activating protein 45 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92619 | |
ARHGAP45 | |
Recombinant protein corresponding to Human HMHA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only