missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HMOX1 (Human) Recombinant Protein

Human HMOX1 full-length ORF ( AAH01491, 1 a.a. - 288 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00003162-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136251

  • 6845.00 SEK / 25µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MERPQPDSMPQDLSEALKEATKEVHTQAENAEFMRNFQKGQVTRDGFKLVMASLYHIYVALEEEIERNKESPVFAPVYFPEELHRKAALEQDLAFWYGPRWQEVIPYTPAMQHYVKRLHEVGRTEPELLVAHAYTRYLGDLSGGQVLKKIAQKALDLPSSGEGLAFFTFPNIASATKFKQLYRSRMNSLEMTPAVRQRVIEEAKTAFLLNIQLFEELQELLTHDTKDQSPSRAPGLRQRASNKVQDSAPVETPRGKPPLNTRSQAPLLRWVLTLSFLVATVAVGLYAM
Specifications

Specifications

AAH01491
heme oxygenase (decycling) 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HMOX1
GST
3162
Wheat germ expression system
25 μg
HO-1, HSP32, bK286B10
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers