missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17674-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
HNRPH3 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two repeats of quasi-RRM domains that bind to RNAs. It is localized in nuclear bodies of the nucleus. This protein is involved in the splicing process and it also participates in early heat shock-induced splicing arrest by transiently leaving the hnRNP complexes.Specifications
HNRNPH3 | |
Polyclonal | |
Unconjugated | |
HNRNPH3 | |
2H9, hnRNP 2H9, hnRNP H3, HNRNPH3, HNRPH3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3189, 361838, 432467 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31942 | |
HNRNPH3 | |
HNRNPH3 Fusion Protein Ag11611 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |