missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPUL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10578-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody can recognize isoform 1-4.
HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55 kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55 kDa in adenovirus-infected cells. This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55 kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55 kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.Specifications
HNRNPUL1 | |
Polyclonal | |
Unconjugated | |
HNRNPUL1 | |
E1B AP5, E1BAP5, HNRNPUL1, HNRPUL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11100, 232989, 361522 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8VDM6, Q9BUJ2 | |
Hnrnpul1 | |
HNRNPUL1 Fusion Protein Ag0899 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |