Läs mer
Invitrogen™ HO-1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142955
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
Specifikationer
| HO-1 | |
| Polyclonal | |
| Unconjugated | |
| HMOX1 | |
| bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3162 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P09601 | |
| HMOX1 | |
| Peptide with sequence C-NKVQDSAPVETPRGK. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |