Läs mer
MedChemExpress HOE-S 785026
Produktkod. 16492717
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Unit Size:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
16492717 50 mg 50mg
16466806 100 mg -
16466628 10 mg -
16409197 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16492717 Supplier MedChemExpress Supplier No. HY15561
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
MedChemExpress HOE-S 785026 is a blue fluorescent dyes, which can be used as a cell dye for DNA.
Specifications
Innehåll och lagring 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Kvantitet 50 mg
Frakt skick Room temperature
Löslighet DMSO : ≥ 37 mg/mL (87.16 mM)
Molekylvikt (g/mol) 424.5 g/mol
CAS 132869-83-1
Synonym meta-Hoechst