Läs mer
MedChemExpress Hoechst 33258 analog 2
Produktkod. p-200104472
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16486846 100 mg 100mg
16472777 50 mg -
16456698 10 mg -
16429257 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16486846 Leverantör MedChemExpress Leverantörsnummer HY15624
MedChemExpress Hoechst 33258 analog 2 are part of a family of blue fluorescent dyes used to stain DNA.
Specifikationer
Innehåll och lagring 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Kvantitet 100 mg
Frakt skick Room temperature
Löslighet H2O : < 0.1 mg/mL (insoluble)
Molekylvikt (g/mol) 422.52 g/mol
CAS 23491-54-5