Learn More
Hoefer™ Clamp Thumbscrews
Accessories for Vertical Protein Electrophoresis Systems.
Brand: Hoefer™ SE6003U
Additional Details : Weight : 0.80000kg
Description
- 12 Pk.
- For 8 cm and 16 cm clamp assemblies
Specifications
Vertical Gel Electrophoresis Unit | |
Clamp Thumbscrew |