Learn More
Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Brand: Hoefer™ HE91-BK
Additional Details : Weight : 0.80000kg
Requires : Comb back must be ordered separately (see ordering table)
Description
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Combs can be used interchangeably with a single comb back.
Specifications
HE99X Max Submarine Electrophoresis Unit | |
2 Screws | |
0.5 cm |
Comb Back | |
0.19 in. |