Learn More
Hoefer™ Gel Casting Accessories for HE 99X Max Submarine Units: Casting Kits
For use with Hoefer HE 99X Max Submarine Electrophoresis Units
Brand:  Hoefer™ HE97X-15
Additional Details : Weight : 0.21600kg
 View more versions of this product
Product Code. 12094837
  • 1698.86 SEK / 1 set
Estimated Shipment: 17-09-2021
Add to basket
Description
Description
For use with Hoefer HE 99X Max Submarine Electrophoresis Units
  • Casting kits include necessary components to cast agarose gels without tape—includes casting tray, running tray, and foam gaskets
Specifications
Specifications
5.9 x 5.9 in. (15 x 15 cm)
Casting Kit
Running tray, casting tray, Foam gasket
15 cm
15 cm
HE99X Max Submarine Electrophoresis Unit
15 x 15 cm
5.9 in.
5.9 in.