Learn More
Hoefer™ HE-PLUS Gel Trays
For use with HE-PLUS Horizontal Electrophoresis System
Brand:  Hoefer™ HE-PLUS13
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 15598921
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Comes in mini (6 x 6 cm), medium (12.5 x 6 cm) or large (12.5 x 13 cm) sizes
  • Either 2 or 4 pcs
Specifications
Specifications
Mini Gel Tray
Gel Tray
4.9 in.
2.36 in.
HE-plus Horizontal Electrophoresis System
4.9 x 2.36 in. (12.5 x 6 cm)
12.5 cm
6 cm