Learn More
Hoefer™ Slotted Gaskets for SE 400 and SE 600 Series Units
Slotted Gaskets for SE 400 and SE 600 Series Units
Brand: Hoefer™ SE4008B
Additional Details : Weight : 0.80000kg
Description
- For use with SE 400 and SE 600 Series Units
Specifications
Gray | |
Slotted Gasket | |
1/Pk. |
SE 400, SE 600 Series Vertical Electrophoresis Unit | |
Silicone Rubber |