Learn More
Hoefer™ SUBHT High Throughput Submarine Gel Unit
Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCR
324.00 SEK - 6925.00 SEK
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12635826
|
Hoefer™
SUB-LEAD |
Replacement High Voltage Lead |
356.25 SEK
Each |
In Stock
Log in to see stock. |
|||||
12675816
|
Hoefer™
SUBHT-CS |
Colored Loading Strip |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12655816
|
Hoefer™
SUBHT |
High Throughput Submarine Gel Unit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12615826
|
Hoefer™
SUBHT-TANK |
Buffer Chamber Assembly |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12655826
|
Hoefer™
SUB-SG |
Silicone Gasket |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12665816
|
Hoefer™
SUBHT-CG |
Casting Gate |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12625826
|
Hoefer™
SUBHT-UT |
Running Tray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12695816
|
Hoefer™
SUBHT-LID |
Lid |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12605826
|
Hoefer™
SUBHT-LUT |
Labeled Running Tray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
- Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
- High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
- Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
- Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
- Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
- This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
- Gel Dimensions: 15 x 15 cm
- Maximum Buffer Volume: 1.2L
- Maximum Power Settings: 300V, 200mA, 60W
- Maximum Sample Capacity: 120
- Maximum Running Temperature: 45°C
- Indoor Use: 4-40°C
- Humidity: up to 80%
- Unit Dimensions: 33.5L x 21.5W x 7 cmH
- Safety Certifications: EN61010-1, CE
Applications:
Single Strand Polymorphism (SSP) analysis
Specifications
SUBHT High Throughput Submarine Gel Unit |