Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Mylar Masks
For use with TE70X, TE77X, TE70XP and TE77XP Semi-Dry Transfer Units
Brand: Hoefer™ TE78
Additional Details : Weight : 0.80000kg
Description
- Comes in regular (16.5 x 18.5 cm) and large (23 x 27.5 cm) sizes
- 4 pcs
Specifications
TE 77 Semi Dry Transfer Unit | |
Mylar™ Mask |