missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Holmium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Holmium(III) fluoride, anhydrous is used to produce the strongest artificially- generated magnetic fields, nuclear control rods, yellow-colored glasses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1143°C |
Physical Form | Powder |
Quantity | 2 g |
UN Number | UN3288 |
Solubility Information | Insoluble in water. |
Formula Weight | 221.93 |
Percent Purity | 99.9% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 7.6 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'