missing translation for 'onlineSavingsMsg'
Learn More

Holmium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7080014
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7440-60-0
Ho
164.93
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
23988
CHEBI:49648
holmium
[Ho]
This item is not returnable. View return policy
7440-60-0
Ho
164.93
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
23988
CHEBI:49648
holmium
[Ho]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Silver
UN Number UN3089
Quantity 5 g
Percent Purity 99.9%
Mesh Size −200 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Holmium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'