Läs mer
Homatropine Bromide, MedChemExpress
Produktkod. 16470988
Encompass_Preferred
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16470988 5 g
16432229 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16470988 Leverantör Medchem Express Leverantörsnummer HYB0547A
MedChemExpress Homatropine Bromide is muscarinic AChR antagonist that is an anticholinergic medication.
Homatropine Bromide is muscarinic AChR antagonist that is an anticholinergic medication.
Specifikationer
Kemiskt namn eller material Homatropine Bromide
Fysisk form Solid
CAS 51-56-9
Färg White
Kvantitet 5 g
Anteckningar om renhetsgrad Research
Molekylformel C16H22BrNO3
För användning med (applikation) Neuroscience-Neuromodulation
Synonym Homatropine hydrobromide
Löslighetsinformation DMSO : 100 mg/mL (280.70 mM; Need ultrasonic) ∣H2O : 100 mg/mL (280.70 mM; Need ultrasonic)
LEDER O=C(C(C1=CC=CC=C1)O)O[C@H]2C[C@@H](CC3)N(C)[C@@H]3C2.Br
Molekylvikt (g/mol) 356.25
Formel vikt 356.25
Procent renhet 99.99%