Läs mer
Homatropine Hydrochloride, LoGiCal
Specifikationer
Specifikationer
| Kemiskt namn eller material | Homatropine Hydrochloride |
| CAS | 637-21-8 |
| Analyt- eller komponentnamn | Homatropine Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C16 H21 N O3 . Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Homatropine hydrochloride (6CI), Benzeneacetic acid, alpha-hydroxy-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (1:1), 1alphaH,5alphaH-Tropan-3alpha-ol, mandelate (ester), hydrochloride (8CI), Benzeneacetic acid, alpha-hydroxy-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (9CI), Benzeneacetic acid, alpha-hydroxy-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride, endo-(+/-)-, Benzeneacetic acid, alpha-hydroxy-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride, endo- |
| InChI formel | InChI=1S/C16H21NO3.ClH/c1-17-12-7-8-13(17)10-14(9-12)20-16(19)15(18)11-5-3-2-4-6-11;/h2-6,12-15,18H,7-10H2,1H3;1H/t12-,13+,14+,15-;/m1./s1 |
| Lösningstyp | Neat |
| LEDER | Cl.CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](O)c3ccccc3 |
| Visa mer |