missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMER2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HOMER2 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828023
|
Proteintech
11143-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818023
|
Proteintech
11143-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function. The encoded protein is a postsynaptic density scaffolding protein. Alternative splicing results in multiple transcript variants. Two related pseudogenes have been identified on chromosome 14.Specifications
HOMER2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9NSB8 | |
9455 | |
HOMER2 Fusion Protein Ag1606 | |
Primary | |
-20°C | |
HOMER2 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACPD, CPD, Cupidin, HOMER 2, homer homolog 2 (Drosophila), Homer protein homolog 2, HOMER2, HOMER2A, HOMER2B, Vesl 2 | |
HOMER2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |