Läs mer
Homogentisic Acid-13C6, TRC
Produktkod. 30008190
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008190 1 mg
30010484 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008190 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH5934021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1216468-48-2
Molekylformel C213C6H8O4
Molekylvikt (g/mol) 174.1
Synonym 2,5-Dihydroxybenzeneacetic Acid-13C6, 2,5-Dihydroxy-α-toluic Acid-13C6, 2,5-Dihydroxyphenylacetic Acid-13C6, Alcapton-13C6, Homogentisinic Acid-13C6, NSC 88940-13C6;
IUPAC-namn 2-(2,5-dihydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)acetic acid
LEDER O[13C]1=[13CH][13CH]=[13C](O)[13C](CC(O)=O)=[13CH]1
Specifikationer
Kemiskt namn eller material Homogentisic Acid-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C8H8O4/c9-6-1-2-7(10)5(3-6)4-8(11)12/h1-3,9-10H,4H2,(H,11,12)/i1+1,2+1,3+1,5+1,6+1,7+1
Formel vikt 174.0624
Procent renhet >95%