Learn More
ATAGO™ Honey Refractometer HHR-2N
Product Code. 10051002
Change view
Click to view available options
Dimensions (L x W x H):
40 x 40 x 170 mm
Unit Size:
Each
Product Code. Dimensions (L x W x H) Unit Size
10051002 40 x 40 x 170 mm Each
1 options
Product Code. 10051002 Supplier ATAGO™ Supplier No. 2522
  • ONLINE EXCLUSIVE PRICE
    2405.00 SEK
    1958.73 SEK / Paket med 1
    Save 446.27 SEK
    18% Av
Beräknad leverans: 30-12-2026
The Honey Refractometer HHR-2N is specially designed to determine the percentage of water contained in honey by means of the refractive index.
  • This handheld honey moisture refractometer features manual temperature compensation.
  • Features:
    • Handheld portable honey refractometer to measure the percentage of water contained in honey (12% to 30%) with a refractive index measurement
    • Minimum water content detection scale of 0.1%
    • Ridged outer shell with a telescope-shaped, streamlined design for gripping, and a spoon-shaped tip for smooth sampling
    • Durable, die-cast aluminum metal body for heat and organic solvent resistance
Specifications
Type Portable Refractometer
Scale Range 12.0% to 30.0%
Model HHR-2N
Resolution 0.1%
Scale Type Brix
Weight (Metric) 260 g
Dimensions (L x W x H) 40 x 40 x 170 mm