Läs mer
Sperian™ Honeywell™ 2000 Comfort-seriens andningsskydd
Produktkod. 15371141
Modell:
2110 FFP1
2110 FFP2
2211 FFP2V
Förpackningsstorlek:
Paket med 20
Paket med 25
Produktkod. 15371141
Brand: Sperian™ 1031593
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
With an ergonomic design to deliver outstanding levels of comfort and adjustment. Sperian™ Honeywell™ 2000 Comfort Series Respirators have a cut-out shape to provide excellent compatibility with goggles.
  • Folds flat packed in individually sealed bags for transportation and storage
  • Dolomite tested, demonstrating the respirators' resistance to clogging over time
  • With high efficiency exhalation valve for protection
  • Double layer face seal provides comfort and protection
TRUSTED_SUSTAINABILITY
Specifikationer
Certifikat och efterlevnad EN 149 : 2001 A1: 2009
Disponibel Single-use
Filtertyp FFP2
Produkttyp Respirator Protection
Storlek Medium
Med utandningsventil Yes
För användning med (applikation) Agriculture, Automotive and Part Manufacturer, Building and Construction, Catering, Chemical Industries, Food Industries, Foundry, General Industry, Glass Industries, Green Spaces, Iron and Steel Industry Laboratory, Maintenance Medical and Pharmaceutical
Latexfri Yes
Modell 2211 FFP2V
Kvantitet 20/Pk.
Form Flat Fold