Learn More
Sperian™ Honeywell™ Premium Disposable Respirators
Highly comfortable and ergonomically designed single-use respirators with no restriction with regard to use. Sperian™ Honeywell™ Premium Disposable Respirators leverage the Willtech™ soft seal technology for an improved fit and exceptional comfort.
Brand: Sperian™ 1005586
Additional Details : Weight : 0.50000kg
Description
- Rigid shell so shape is maintained even in moist and heavy loading environments
- Hyopallergenic flange seal with absorption properties
- Preformed and colored nose bridge to save time
- Large internal volume to easily communicate and breathe
- Twin elastic straps, ultra flexible and resistant, stapled outside of the filtering area
- No latex, no silicone, no PVC
- Protection and wearer acceptance guaranteed during periods of extended use
Specifications
EN 149:2001A1:2009, ISO 9001 / 2000, EC | |
FFP2 | |
Medium | |
Textile, mining, construction, iron and steel industries, car body preparation industry, wood working, agricultural and horticultural activities, food industry, cutting and molding of Metals etc | |
5211 ML Respirator | |
Dusts, Mists, Fumes |
Single-use | |
Respiratory Protection | |
Yes | |
Yes | |
20/Pk. | |
Molded Cup |