Learn More
Sperian™ Honeywell™ A2 Organic Vapour Filter Canisters for Respirators
Perfect shape to allow great vision in all situations. Sperian™ Honeywell™ A2 Organic Vapour Filter Canisters for Respirators can be used in places where the oxygen concentration is safely upper 17%.
Brand: Sperian™ 1001577
Additional Details : Weight : 0.86400kg
Description
- Can only be used with a half maks Valuair™, Premier™ or MX/PF or a full face Optifit Twin™ equipped with a T system
- User checks installation via the audible click on its safety system
- Coconut-based carbon charcoal impregnated for a high quality and safe level of filtration
- Mechanical filtration membrane
- Housing made of solid and resistant plastic material