Learn More
Sperian™ Honeywell™ Metal Hearing Protection
Click to view available options
Color:
Black
Black (Headband), Dark Gray (Cup)
Description
- Designed to fit and seal to the workers ear perfectly
- Reduces background noises to safe levels
- Efficiently collect noise exposure, helping identify risks and take action
Specifications
Color | Black (Headband), Dark Gray (Cup) |
Headband Material | Steel |
For Use With (Application) | To block noise |
Product Type | Hearing Protection |
Headband Color | Black |
Material | Metal |