missing translation for 'onlineSavingsMsg'
Learn More

HOP2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11339-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888343

  • 1355.00 SEK / 20µL
Estimated Shipment: 28-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PSMC3IP plays an important role in meiotic recombination. It stimulates DMC1-mediated strand exchange required for pairing homologous chromosomes during meiosis. The complex PSMC3IP/MND1 binds DNA, stimulates the recombinase activity of DMC1 as well as DMC1 D-loop formation from double-strand DNA. This complex stabilizes presynaptic RAD51 and DMC1 filaments formed on single strand DNA to capture double-strand DNA. This complex stimulates both synaptic and presynaptic critical steps in RAD51 and DMC1-promoted homologous pairing. It may inhibit HIV-1 viral protein TAT activity and modulate the activity of proteasomes through association with PSMC3.
Specifications

Specifications

HOP2
Polyclonal
Unconjugated
PSMC3IP
GT198, HOP2, HUMGT198A, PSMC3 interacting protein, PSMC3IP, TBP 1 interacting protein, TBPIP
Rabbit
Antigen Affinity Chromatography
RUO
29893
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9P2W1
PSMC3IP
HOP2 Fusion Protein Ag1883
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers