Läs mer
Hopantenate Calcium, TRC
Produktkod. 30011389
Change view
Kvantitet:
1 mg
2.5 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011389 1 mg -
30009267 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011389 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH6695651MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17097-76-6
Molekylformel 2(C20H19NO5) • Ca
Molekylvikt (g/mol) 504.59
Synonym Calcium bis[4-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]butanoate], Calcium hopantenate, Calcium hopantenate anhydrous
IUPAC-namn calcium;4-[[rac-(2R)-2,4-dihydroxy-3,3-dimethyl-butanoyl]amino]butanoate
LEDER [Ca+2].CC(C)(CO)[C@@H](O)C(=O)NCCCC(=O)[O-].CC(C)(CO)[C@@H](O)C(=O)NCCCC(=O)[O-]
Specifikationer
Kemiskt namn eller material Hopantenate calcium salt
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/2C10H19NO5.Ca.H2O/c2*1-10(2,6-12)8(15)9(16)11-5-3-4-7(13)14;;/h2*8,12,15H,3-6H2,1-2H3,(H,11,16)(H,13,14);;1H2/q;;+2;/p-2/t2*8-;;/m00../s1
Kvantitet 1 mg
Formel vikt 504.1996
Procent renhet >95%