Läs mer
Hordenine Hydrochloride (4-Hydroxy-N,N-dimethylphenethylamine Hydrochloride) 1.0 mg/ml in Methanol (as free base), LoGiCal
Produktkod. 30458266
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30458266 1 ml
1 options
Produktkod. 30458266 Leverantör LoGiCal Leverantörsnummer LGCAMP1275.4901
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hordenine Hydrochloride (4-Hydroxy-N,N-dimethylphenethylamine Hydrochloride)
CAS 6027-23-2
Analyt- eller komponentnamn Hordenine Hydrochloride (4-Hydroxy-N,N-dimethylphenethylamine Hydrochloride)
Frakt skick Ice pack (-20°C)
Molekylformel C10 H15 N O . Cl H
Certifikat och efterlevnad ISO 17034
Synonym Hordenine Hydrochloride, 4-Hydroxy-N,N-dimethylphenethylamine Hydrochloride
InChI formel InChI=1S/C10H15NO.ClH/c1-11(2)8-7-9-3-5-10(12)6-4-9;/h3-6,12H,7-8H2,1-2H3;1H
Lösningstyp Single Solution
LEDER Cl.CN(C)CCc1ccc(O)cc1
IUPAC-namn 4-[2-(dimethylamino)ethyl]phenol;hydrochloride
Lösningsmedel eller matris Methanol
Industry Type Clinical Testing
Molekylvikt (g/mol) 201.69
Kvantitet 1 ml
Formel vikt 201.092 g/mol