Learn More
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DB Series
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DB Series is designed for simple process applications and has no options available for process optimization.
POA
Specifications
Product Type Horizontal, Front-loading Benchtop Autoclave
View More Specs
Description
Description
  • Standard features:
    • Internal heating elements within the autoclave chamber
    • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
    • Temperature and pressure range 140°C, 4 bar (except for Systec DB-23)
    • LCD display and fully automatic microprocessor control
    • Number of sterilization programs: 3
    • Code-secured access rights for changing parameters and further safety-relevant intervention
    • Flexible PT-100 temperature sensor
  • Optional: Integrated printer for batch documentation
Specifications
Specifications
Horizontal, Front-loading Benchtop Autoclave