Learn More
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DB Series
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DB Series is designed for simple process applications and has no options available for process optimization.
POA
Description
- Standard features:
- Internal heating elements within the autoclave chamber
- Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
- Temperature and pressure range 140°C, 4 bar (except for Systec DB-23)
- LCD display and fully automatic microprocessor control
- Number of sterilization programs: 3
- Code-secured access rights for changing parameters and further safety-relevant intervention
- Flexible PT-100 temperature sensor
- Optional: Integrated printer for batch documentation
Specifications
Horizontal, Front-loading Benchtop Autoclave |